Learn More
Memmert™ 4 - 20 mA Current Loop Interface O2 Controller
4-20mA current loop interface for O2 control option
Brand:  Memmert™ ICO150V1
 View more versions of this product
Product Code. 17109974
  • 1815.00 DKK / Each
Only null left
Estimated Shipment: 28-08-2025
Description
  • Requires option T6
  • 0 to 25 % O2 = 4 - 20 mA
Specifications
Specifications
Requires option T6
ICO150
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ 4 - 20 mA Current Loop Interface O2 Controller > ICO150