missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ 4 - 20 mA Current Loop Interface

4-20mA current loop interface Humidity controller 0-100% RH, actual value

Brand:  Memmert™ ICO50V7

 View more versions of this product

Product Code. 17190035

  • 1815.00 DKK / Each
Only null left
Estimated Shipment: 29-08-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Requires option K7 or K6
  • Humidity controller
  • Actual value (0 to 100 % rh = 4 - 20 mA)
Specifications

Specifications

Requires option K7 or K6
ICO50
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Memmert™ 4 - 20 mA Current Loop Interface > ICO50

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.