Learn More
Alfa Aesar™ 4-(2-Thienyl)benzoic acid, 96%
Brand: Alfa Aesar™ H33642.03
Additional Details : CAS Number : 29886-62-2 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
29886-62-2 | |
204.243 | |
CVDUBQJEQNRCIZ-UHFFFAOYSA-N | |
736498 | |
C1=CSC(=C1)C2=CC=C(C=C2)C(=O)O |
C11H8O2S | |
MFCD00690742 | |
4-thiophen-2-yl benzoic acid, 4-2-thienyl benzoic acid, 4-thiophen-2-yl-benzoic acid, benzoic acid,4-2-thienyl, benzoic acid, 4-2-thienyl, pubchem12353, acmc-1cb4z, 4-2-thienyl benzoicacid, 4-thien-2-ylbenzoic acid, amtda058 | |
4-thiophen-2-ylbenzoic acid |
Specifications
4-(2-Thienyl)benzoic acid | |
1g | |
C11H8O2S | |
4-thiophen-2-yl benzoic acid, 4-2-thienyl benzoic acid, 4-thiophen-2-yl-benzoic acid, benzoic acid,4-2-thienyl, benzoic acid, 4-2-thienyl, pubchem12353, acmc-1cb4z, 4-2-thienyl benzoicacid, 4-thien-2-ylbenzoic acid, amtda058 | |
C1=CSC(=C1)C2=CC=C(C=C2)C(=O)O | |
204.243 | |
204.25 |
243°C to 245°C | |
29886-62-2 | |
MFCD00690742 | |
CVDUBQJEQNRCIZ-UHFFFAOYSA-N | |
4-thiophen-2-ylbenzoic acid | |
736498 | |
96% |