Få mere at vide
4-(2-Methylpropyl)piperidine-2,6-dione (3-Isobutylglutarimide), Mikromol™
Artikelnummer. 30460651
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30460651 4 x 25 mg
30049764 25 mg
17941116 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30460651 Leverandør Mikromol Leverandørnr. MM1376.1900254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(2-Methylpropyl)piperidine-2,6-dione (3-Isobutylglutarimide), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(2-Methylpropyl)piperidine-2,6-dione
CAS 916982-10-0
Analyt- eller komponentnavne 4-(2-Methylpropyl)piperidine-2,6-dione
Forsendelsestilstand Room Temperature
Molekylær formel C9 H15 N O2
Certificeringer/compliance ISO 9001
Synonym 2,6-Piperidinedione, 4-(2-methylpropyl)-, -Isobutylglutarimide, 4-(2-Methylpropyl)piperidine-2,6-dione, 4-Isobutyl-2,6-dioxopiperidine, 4-Isobutylpiperidine-2,6-dione
InChI formel InChI=1S/C9H15NO2/c1-6(2)3-7-4-8(11)10-9(12)5-7/h6-7H,3-5H2,1-2H3,(H,10,11,12)
Løsningstype Neat
SMIL CC(C)CC1CC(=O)NC(=O)C1
IUPAC navn 4-(2-methylpropyl)piperidine-2,6-dione
Molekylvægt (g/mol) 169.22
Mængde 4 x 25 mg
Formel vægt 169.1103 g/mol