missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methyl-benzoimidazol-1-yl)-butanoic acid 97+%, Thermo Scientific™

Catalog Number p-4510296
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
115444-73-0
C12H14N2O2
218.26
MFCD00879013
IRBRKNDBYUBKIR-UHFFFAOYSA-N
4-2-methyl-1h-benzimidazol-1-yl butanoic acid, 4-2-methyl-benzoimidazol-1-yl-butyric acid, 1h-benzimidazole-1-butanoicacid, 2-methyl, 4-2-methylbenzimidazol-1-yl butanoic acid, 4-2-methyl-1,3-benzodiazol-1-yl butanoic acid, 4-2-methylbenzimidazolyl butanoic acid, acmc-1c9be, 1-3-carboxypropyl-2-methylbenzimidazole, 4-2-methyl-1-benzimidazolyl butanoic acid, 4-2-methyl-1h-benzo d imidazol-1-yl butanoic acid
4379684
4-(2-methylbenzimidazol-1-yl)butanoic acid
CC1=NC2=CC=CC=C2N1CCCC(O)=O
This item is not returnable. View return policy
115444-73-0
C12H14N2O2
218.26
MFCD00879013
IRBRKNDBYUBKIR-UHFFFAOYSA-N
4-2-methyl-1h-benzimidazol-1-yl butanoic acid, 4-2-methyl-benzoimidazol-1-yl-butyric acid, 1h-benzimidazole-1-butanoicacid, 2-methyl, 4-2-methylbenzimidazol-1-yl butanoic acid, 4-2-methyl-1,3-benzodiazol-1-yl butanoic acid, 4-2-methylbenzimidazolyl butanoic acid, acmc-1c9be, 1-3-carboxypropyl-2-methylbenzimidazole, 4-2-methyl-1-benzimidazolyl butanoic acid, 4-2-methyl-1h-benzo d imidazol-1-yl butanoic acid
4379684
4-(2-methylbenzimidazol-1-yl)butanoic acid
CC1=NC2=CC=CC=C2N1CCCC(O)=O
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 218.25
Percent Purity 90%
Chemical Name or Material 4-(2-Methyl-1H-benzimidazol-1-yl)butanoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'