Få mere at vide
4-(2-Hydroxyethyl-d4)morpholine, CDN
Artikelnummer. 30320193
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320193 0.1 g 0.1g
30312786 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320193 Leverandør CDN Leverandørnr. CDND66100.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Morpholineethanol-d4
CAS 1185052-90-7
Molekylær formel C6 2H4 H9 N O2
Mængde 0.1 g
InChI formel InChI=1 S/C6H13NO2/c8-4-1-7-2-5-9-6-3-7/h8H,1-6H2/i1D2,4D2
SMIL [2 H]C([2 H])(O)C([2 H])([2 H])N1CCOCC1
IUPAC navn 1,1,2,2-tetradeuterio-2-morpholin-4-ylethanol
Molekylvægt (g/mol) 135.2
Formel vægt 135.1197 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity