Få mere at vide
4-[[2-Ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]benzoyl]amino]-1-methyl-3-propyl-1H-pyrazole-5-carboxamide, Mikromol™
Artikelnummer. 30049767
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049767 25 mg
30050067 100 mg
30458797 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30049767 Leverandør Mikromol Leverandørnr. MM0254.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-[[2-Ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]benzoyl]amino]-1-methyl-3-propyl-1H-pyrazole-5-carboxamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-[(2-Ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]benzoyl)amino]-1-methyl-3-propyl-1H-pyrazole-5-carboxamide
CAS 200575-15-1
Analyt- eller komponentnavne 4-[(2-Ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]benzoyl)amino]-1-methyl-3-propyl-1H-pyrazole-5-carboxamide
Forsendelsestilstand Room Temperature
Molekylær formel C22 H32 N6 O5 S
Certificeringer/compliance ISO 17025
Synonym 4-[[2-Ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]benzoyl]amino]-1-methyl-3-propyl-1H-pyrazole-5-carboxamide
InChI formel InChI=1S/C22H32N6O5S/c1-5-7-17-19(20(21(23)29)27(4)25-17)24-22(30)16-14-15(8-9-18(16)33-6-2)34(31,32)28-12-10-26(3)11-13-28/h8-9,14H,5-7,10-13H2,1-4H3,(H2,23,29)(H,24,30)
SMIL CCCc1nn(C)c(C(=O)N)c1NC(=O)c2cc(ccc2OCC)S(=O)(=O)N3CCN(C)CC3
IUPAC navn 4-[[2-ethoxy-5-(4-methylpiperazin-1-yl)sulfonylbenzoyl]amino]-2-methyl-5-propylpyrazole-3-carboxamide
Molekylvægt (g/mol) 492.59
Mængde 25 mg
Formel vægt 492.2155 g/mol