Få mere at vide
4-[2-(Dipropylamino)ethyl]indoline-2,3-dione Hydrochloride, Mikromol™
Beskrivelse
4-[2-(Dipropylamino)ethyl]indoline-2,3-dione Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-Oxo Ropinirole hydrochloride |
| CAS | 221264-21-7 |
| Analyt- eller komponentnavne | 3-Oxo Ropinirole hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H22 N2 O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 1H-Indole-2,3-dione, 4-[2-(dipropylamino)ethyl]-, hydrochloride (1:1), 1H-Indole-2,3-dione, 4-[2-(dipropylamino)ethyl]-, monohydrochloride (9CI), SKF-96266-A, Ph Eur Ropinirole Impurity A, USP Ropinirole Related Compound B, SKF-96266, SB-270341, RRT = 1.4 [SB-410189], SKF-98959, RRT = 28.5 [SB-381331], SKF-96413, Ropinirole Related Compound B as Hydrochloride, Ropinirole Hydrochloride Impurity A as Hydrochloride, Ropinirole Impurity A as Hydrochloride |
| InChI formel | InChI=1S/C16H22N2O2.ClH/c1-3-9-18(10-4-2)11-8-12-6-5-7-13-14(12)15(19)16(20)17-13;/h5-7H,3-4,8-11H2,1-2H3,(H,17,19,20);1H |
| SMIL | Cl.CCCN(CCC)CCc1cccc2NC(=O)C(=O)c12 |
| IUPAC navn | 4-[2-(dipropylamino)ethyl]-1H-indole-2,3-dione;hydrochloride |
| Vis mere |