missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloroacetyl)-2H-1,4-benzothiazine, 97%, Thermo Scientific™

Catalog Number p-7041138
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
20751-75-1
C10H10ClNOS
227.706
MFCD06655687
AYFZGNOUGDUMDS-UHFFFAOYSA-N
4-chloroacetyl-3,4-dihydro-2h-1,4-benzothiazine, 4-2-chloroacetyl-2h-1,4-benzothiazine, 2-chloro-1-3,4-dihydro-2h-1,4-benzothiazin-4-yl ethan-1-one, 2-chloro-1-2,3-dihydro-1,4-benzothiazin-4-yl ethanone, ethanone, 2-chloro-1-2,3-dihydro-4h-1,4-benzothiazin-4-yl, 2-chloro-1-10-thia-7-azabicyclo 4.4.0 deca-1,3,5-trien-7-yl ethanone, 1-2h-benzo b 1,4 thiazin-4 3h-yl-2-chloroethanone, 2-chloro-1-2,3-dihydro-4h-1,4-benzothiazin-4-yl ethan-1-one
4962259
2-chloro-1-(2,3-dihydro-1,4-benzothiazin-4-yl)ethanone
C1CSC2=CC=CC=C2N1C(=O)CCl
This item is not returnable. View return policy
20751-75-1
C10H10ClNOS
227.706
MFCD06655687
AYFZGNOUGDUMDS-UHFFFAOYSA-N
4-chloroacetyl-3,4-dihydro-2h-1,4-benzothiazine, 4-2-chloroacetyl-2h-1,4-benzothiazine, 2-chloro-1-3,4-dihydro-2h-1,4-benzothiazin-4-yl ethan-1-one, 2-chloro-1-2,3-dihydro-1,4-benzothiazin-4-yl ethanone, ethanone, 2-chloro-1-2,3-dihydro-4h-1,4-benzothiazin-4-yl, 2-chloro-1-10-thia-7-azabicyclo 4.4.0 deca-1,3,5-trien-7-yl ethanone, 1-2h-benzo b 1,4 thiazin-4 3h-yl-2-chloroethanone, 2-chloro-1-2,3-dihydro-4h-1,4-benzothiazin-4-yl ethan-1-one
4962259
2-chloro-1-(2,3-dihydro-1,4-benzothiazin-4-yl)ethanone
C1CSC2=CC=CC=C2N1C(=O)CCl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 227.71
Percent Purity 97%
Chemical Name or Material 4-(2-Chloroacetyl)-2H-1,4-benzothiazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'