Få mere at vide
4-(2-Bromophenyl)-2,6-dimethyl-3,5-pyridinedicarboxylic Acid-d10 Diethyl Ester, TRC
Artikelnummer. 30341583
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30341583 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341583 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6864971MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(2-Bromophenyl)-2,6-dimethyl-3,5-pyridinedicarboxylic Acid-d10 Diethyl Ester
CAS 1329793-25-0
Molekylær formel C19H12D10BrNO4
Mængde 1 mg
Synonym 4-(2-Bromophenyl)-2,6-dimethyl-3,5-pyridinedicarboxylic Acid-d10 3,5-Diethyl Ester
InChI formel InChI=1S/C19H22BrNO4/c1-5-24-18(22)15-11(3)21-12(4)16(19(23)25-6-2)17(15)13-9-7-8-10-14(13)20/h7-10,17,21H,5-6H2,1-4H3/i1D3,2D3,5D2,6D2
SMIL [2H]C([2H])([2H])C([2H])([2H])OC(=O)C1=C(C)NC(=C(C1c2ccccc2Br)C(=O)OC([2H])([2H])C([2H])([2H])[2H])C
IUPAC navn bis(1,1,2,2,2-pentadeuterioethyl) 4-(2-bromophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molekylvægt (g/mol) 418.35
Formel vægt 417.136