missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Bromo-6-fluorobenzyl)morpholine, 96%, Thermo Scientific™

Catalog Number 15488778
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1345472-12-9
C11H13BrFNO
274.133
CWQNAYDNRWPWBM-UHFFFAOYSA-N
1-bromo-3-fluoro-2-morpholinomethyl benzene, 4-2-bromo-6-fluorophenyl methyl morpholine, acmc-209bvy
56776947
4-[(2-bromo-6-fluorophenyl)methyl]morpholine
C1COCCN1CC2=C(C=CC=C2Br)F
This item is not returnable. View return policy
1345472-12-9
C11H13BrFNO
274.133
CWQNAYDNRWPWBM-UHFFFAOYSA-N
1-bromo-3-fluoro-2-morpholinomethyl benzene, 4-2-bromo-6-fluorophenyl methyl morpholine, acmc-209bvy
56776947
4-[(2-bromo-6-fluorophenyl)methyl]morpholine
C1COCCN1CC2=C(C=CC=C2Br)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 274.13
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-(2-Bromo-6-fluorobenzyl)morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'