missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Acetoxyphenyl)-2-(Fmoc-amino)-5-methylthiazole, 97%, Thermo Scientific™

Catalog Number 15484676
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
1421262-42-1
C27H22N2O4S
470.543
MFCD22988980
MAUMOHZLUIAQJC-UHFFFAOYSA-N
2-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-methyl-1,3-thiazol-4-yl phenyl acetate, 4-2-acetoxyphenyl-2-fmoc-amino-5-methylthiazole, 2-2-9h-fluoren-9-yl methoxy carbonylamino-5-methylthiazol-4-yl phenyl acetate
71741371
[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methyl-1,3-thiazol-4-yl]phenyl] acetate
CC1=C(N=C(S1)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C5=CC=CC=C5OC(=O)C
This item is not returnable. View return policy
1421262-42-1
C27H22N2O4S
470.543
MFCD22988980
MAUMOHZLUIAQJC-UHFFFAOYSA-N
2-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-methyl-1,3-thiazol-4-yl phenyl acetate, 4-2-acetoxyphenyl-2-fmoc-amino-5-methylthiazole, 2-2-9h-fluoren-9-yl methoxy carbonylamino-5-methylthiazol-4-yl phenyl acetate
71741371
[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methyl-1,3-thiazol-4-yl]phenyl] acetate
CC1=C(N=C(S1)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C5=CC=CC=C5OC(=O)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 2 g
Formula Weight 470.54
Percent Purity 97%
Chemical Name or Material 4-(2-Acetoxyphenyl)-2-(Fmoc-amino)-5-methylthiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'