missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
4-[2’-(7’’-Deazaguanine)ethyl]benzoic Acid N-Hydroxysuccinimide Ester, TRC
Specifications
Specifications
| Kemisk navn eller materiale | 4-[2'-(7''-Deazaguanine)ethyl]benzoic Acid N-Hydroxysuccinimide Ester |
| CAS | 204257-65-8 |
| Molekylær formel | C19 H17 N5 O5 |
| Mængde | 25 mg |
| Synonym | Benzoic acid, 4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]-, 2,5-dioxo-1-pyrrolidinyl ester, 2,5-Pyrrolidinedione, 1-[[4-[2-(2-amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]oxy]- (9CI), 4-[2'-(7''-Deazaguanine)ethyl]benzoic acid N-hydroxysuccinimide ester |
| InChI formel | InChI=1S/C19H17N5O5/c20-19-22-16-15(17(27)23-19)12(9-21-16)6-3-10-1-4-11(5-2-10)18(28)29-24-13(25)7-8-14(24)26/h1-2,4-5,9H,3,6-8H2,(H4,20,21,22,23,27) |
| SMIL | NC1=NC(=O)c2c(CCc3ccc(cc3)C(=O)ON4C(=O)CCC4=O)c[nH]c2N1 |
| IUPAC navn | (2,5-dioxopyrrolidin-1-yl) 4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate |
| Molekylvægt (g/mol) | 395.37 |
| Formel vægt | 395.123 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?