Få mere at vide
4-[2-(5-Ethyl-2-pyridinyl)-d4-ethoxy]benzaldehyde, TRC
Artikelnummer. 30371783
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30371783 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371783 Leverandør Toronto Research Chemicals Leverandørnr. TRCE92558225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-[2-(5-Ethyl-2-pyridinyl)-d4-ethoxy]benzaldehyde
CAS 1189479-80-8
Molekylær formel C16 2H4 H13 N O2
Mængde 25 mg
InChI formel InChI=1S/C16H17NO2/c1-2-13-3-6-15(17-11-13)9-10-19-16-7-4-14(12-18)5-8-16/h3-8,11-12H,2,9-10H2,1H3/i9D2,10D2
SMIL [2H]C([2H])(Oc1ccc(C=O)cc1)C([2H])([2H])c2ccc(CC)cn2
IUPAC navn 4-[1,1,2,2-tetradeuterio-2-(5-ethylpyridin-2-yl)ethoxy]benzaldehyde
Molekylvægt (g/mol) 259.34
Formel vægt 259.151