Få mere at vide
4-(2,4-Dichlorophenoxy-d3)butyric Acid, CDN
Artikelnummer. 30318885
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318885 0.01 g
30316777 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318885 Leverandør CDN Leverandørnr. CDND65850.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4-DB D3 (phenyl-D3)
CAS 1219802-46-6
Molekylær formel C10 D3 H7 Cl2 O3
Mængde 0.01 g
Synonym 4-(2,4-Dichlorophenoxy-d3)butyric Acid, 2,4-DB-d3 (ring-d3), 2,4-DB D3 (phenyl-D3), 4-(2,4-Dichlorophenoxy-3,5,6-d3)butanoic acid
InChI formel InChI=1 S/C10H10Cl2O3/c11-7-3-4-9(8(12)6-7)15-5-1-2-10(13)14/h3-4,6 H,1-2,5H2,(H,13,14)/i3D,4 D,6 D
SMIL [2 H]c1c([2 H])c(OCCCC(=O)O)c(Cl)c([2 H])c1Cl
IUPAC navn 4-(2,4-dichloro-3,5,6-trideuteriophenoxy)butanoic acid
Molekylvægt (g/mol) 252.1091
Formel vægt 251.0195 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity