Få mere at vide
4-((2,4-Dichloro-5-methoxyphenyl)amino)-7-(3-(dimethylamino)propoxy)-6-methoxyquinoline-3-carbonitrile Dimer, TRC
Artikelnummer. 30375593
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30375593 100 mg
30377196 10 mg
30375771 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30375593 Leverandør Toronto Research Chemicals Leverandørnr. TRCD424570100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2095306-26-4
Molekylær formel C46H44Cl4N8O6
Molekylvægt (g/mol) 946.7
Synonym 4-((2,4-Dichloro-5-methoxyphenyl)amino)-7-(3-(4-(3-((4-((2,4-dichloro-5-methoxyphenyl)amino)-3-isocyano-6-methoxyquinolin-7-yl)oxy)propyl)piperazin-1-yl)propoxy)-6-methoxyquinoline-3-carbonitrile, Bosutinib dimer
SMIL COc1cc(Nc2c(cnc3cc(OCCCN4CCN(CCCOc5cc6ncc([N+]#[C-])c(Nc7cc(OC)c(Cl)cc7Cl)c6cc5OC)CC4)c(OC)cc23)C#N)c(Cl)cc1Cl
Tekniske data
Kemisk navn eller materiale 4-((2,4-Dichloro-5-methoxyphenyl)amino)-7-(3-(dimethylamino)propoxy)-6-methoxyquinoline-3-carbonitrile Dimer
Mængde 100 mg
InChI formel InChI=1S/C46H44Cl4N8O6/c1-52-38-26-54-35-21-44(42(62-5)17-29(35)46(38)56-37-23-40(60-3)33(50)19-31(37)48)64-15-7-9-58-12-10-57(11-13-58)8-6-14-63-43-20-34-28(16-41(43)61-4)45(27(24-51)25-53-34)55-36-22-39(59-2)32(49)18-30(36)47/h16-23,25-26H,6-15H2,2-5H3,(H,53,55)(H,54,56)
Formel vægt 944.2138