Få mere at vide
4-(2,4-Diaminophenyl)-3-morpholinone, TRC
Artikelnummer. 30362756
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362756 50 mg
30355466 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30362756 Leverandør Toronto Research Chemicals Leverandørnr. TRCD41661050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 482308-13-4
Molekylær formel C10H13N3O2
Molekylvægt (g/mol) 207.23
SMIL Nc1ccc(N2CCOCC2=O)c(N)c1
Tekniske data
Kemisk navn eller materiale 4-(2,4-Diaminophenyl)-3-morpholinone
Mængde 50 mg
InChI formel InChI=1S/C10H13N3O2/c11-7-1-2-9(8(12)5-7)13-3-4-15-6-10(13)14/h1-2,5H,3-4,6,11-12H2
Formel vægt 207.1008