Få mere at vide
4-[2-(4-Chlorobutylidene)hydrazinyl]-N-methylbenzenemethanesulfonamide, TRC
Artikelnummer. 30359936
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359936 25 mg
30360062 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30359936 Leverandør Toronto Research Chemicals Leverandørnr. TRCC88918025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-[2-(4-Chlorobutylidene)hydrazinyl]-N-methylbenzenemethanesulfonamide
CAS 88918-68-7
Molekylær formel C12 H18 Cl N3 O2 S
Mængde 25 mg
Synonym Benzenemethanesulfonamide, 4-[2-(4-chlorobutylidene)hydrazinyl]-N-methyl-, Benzenemethanesulfonamide, 4-[(4-chlorobutylidene)hydrazino]-N-methyl- (9CI)
InChI formel InChI=1S/C12H18ClN3O2S/c1-14-19(17,18)10-11-4-6-12(7-5-11)16-15-9-3-2-8-13/h4-7,9,14,16H,2-3,8,10H2,1H3
SMIL CNS(=O)(=O)Cc1ccc(NN=CCCCCl)cc1
IUPAC navn 1-[4-[2-(4-chlorobutylidene)hydrazinyl]phenyl]-N-methylmethanesulfonamide
Molekylvægt (g/mol) 303.81
Formel vægt 303.0808