Få mere at vide
4-(2,3-Epoxypropoxy)phenylacetic Acid, Mikromol™
Artikelnummer. 30459126
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459126 4 x 25 mg
30050305 100 mg
30052623 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30459126 Leverandør Mikromol Leverandørnr. MM0013.1100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(2,3-Epoxypropoxy)phenylacetic Acid, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(2,3-Epoxypropoxy)phenylacetic Acid
CAS 72224-25-0
Analyt- eller komponentnavne 4-(2,3-Epoxypropoxy)phenylacetic Acid
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C11 H12 O4
Certificeringer/compliance ISO 9001
Synonym 4-(2-oxiranylmethoxy)-Benzeneacetic acid, Benzeneacetic acid, 4-(oxiranylmethoxy)- (9CI), Benzeneacetic acid, 4-(oxiranylmethoxy)-, (+/-)-
InChI formel InChI=1S/C11H12O4/c12-11(13)5-8-1-3-9(4-2-8)14-6-10-7-15-10/h1-4,10H,5-7H2,(H,12,13)
Løsningstype Neat
SMIL OC(=O)Cc1ccc(OCC2CO2)cc1
IUPAC navn 2-[4-(oxiran-2-ylmethoxy)phenyl]acetic acid
Molekylvægt (g/mol) 208.21
Mængde 4 x 25 mg
Formel vægt 208.0736 g/mol