Få mere at vide
[[4-[[2-(1-Methylethoxy)ethoxy]methyl]phenoxy]methyl]oxirane-d5, TRC
Artikelnummer. 30474821
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30474821 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30474821 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30476710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale ((4-((2-(1-Methylethoxy)ethoxy)methyl)phenoxy)methyl)oxirane-d5
CAS 1215808-65-3
Molekylær formel C15H17D5O4
Mængde 10 mg
Synonym 2-[[4-[[2-(1-Methylethoxy)ethoxy]methyl]phenoxy]methyl]oxirane-d5[[4-[[2-Isopropoxyethoxy]methyl]phenoxy]methyl]oxirane-d5
InChI formel InChI=1S/C15H22O4/c1-12(2)17-8-7-16-9-13-3-5-14(6-4-13)18-10-15-11-19-15/h3-6,12,15H,7-11H2,1-2H3/i10D2,11D2,15D
SMIL [2H]C([2H])(Oc1ccc(COCCOC(C)C)cc1)C2([2H])OC2([2H])[2H]
IUPAC navn 2,2,3-trideuterio-3-[dideuterio-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]methyl]oxirane
Molekylvægt (g/mol) 271.36
Formel vægt 271.1832