Få mere at vide
4-[2-[(1,1-Dimethylethyl)amino]ethyl]-2-methylphenol, Mikromol™
Artikelnummer. 30462526
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462526 4 x 25 mg
30052447 25 mg
30051356 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30462526 Leverandør Mikromol Leverandørnr. MM0381.0800254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-[2-[(1,1-Dimethylethyl)amino]ethyl]-2-methylphenol, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-[2-[(1,1-Dimethylethyl)amino]ethyl]-2-methylphenol
CAS 132183-64-3
Analyt- eller komponentnavne 4-[2-[(1,1-Dimethylethyl)amino]ethyl]-2-methylphenol
Forsendelsestilstand Room Temperature
Molekylær formel C13 H21 N O
Certificeringer/compliance ISO 17025
Synonym 4-[2-[(1,1-Dimethylethyl)amino]ethyl]-2-methylphenol, Salbutamol Impurity H
InChI formel InChI=1S/C13H21NO/c1-10-9-11(5-6-12(10)15)7-8-14-13(2,3)4/h5-6,9,14-15H,7-8H2,1-4H3
Løsningstype Neat
SMIL Cc1cc(CCNC(C)(C)C)ccc1O
IUPAC navn 4-[2-(tert-butylamino)ethyl]-2-methylphenol
Molekylvægt (g/mol) 207.31
Mængde 4 x 25 mg
Formel vægt 207.1623 g/mol