missing translation for 'onlineSavingsMsg'
Få mere at vide

4-[2-[(1,1-Dimethylethyl)amino]ethyl]-2-methylphenol, Mikromol™

Artikelnummer. 30052447
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052447 25 mg
30051356 100 mg
30462526 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30052447 Leverandør Mikromol Leverandørnr. MM0381.080025

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-[2-[(1,1-Dimethylethyl)amino]ethyl]-2-methylphenol
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 4-[2-[(1,1-Dimethylethyl)amino]ethyl]-2-methylphenol
CAS 132183-64-3
Analyt- eller komponentnavne 4-[2-[(1,1-Dimethylethyl)amino]ethyl]-2-methylphenol
Forsendelsestilstand Room Temperature
Molekylær formel C13 H21 N O
Certificeringer/compliance ISO 17025
Synonym 4-[2-[(1,1-Dimethylethyl)amino]ethyl]-2-methylphenol, Salbutamol Impurity H
InChI formel InChI=1S/C13H21NO/c1-10-9-11(5-6-12(10)15)7-8-14-13(2,3)4/h5-6,9,14-15H,7-8H2,1-4H3
SMIL Cc1cc(CCNC(C)(C)C)ccc1O
IUPAC navn 4-[2-(tert-butylamino)ethyl]-2-methylphenol
Molekylvægt (g/mol) 207.31
Mængde 25 mg
Formel vægt 207.1623 g/mol
Vis mere Vis mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.