Få mere at vide
4-[(1S)-4-(Dimethylamino)-1-(4-fluorophenyl)-1-hydroxybutyl]-3-(hydroxymethyl)benzonitrile Hemi-(+)-di-p-toluoyltartrate, Mikromol™
Beskrivelse
4-[(1S)-4-(Dimethylamino)-1-(4-fluorophenyl)-1-hydroxybutyl]-3-(hydroxymethyl)benzonitrile Hemi-(+)-di-p-toluoyltartrate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-((1S)-4-(Dimethylamino)-1-(4-fluorophenyl)-1-hydroxybutyl)-3-(hydroxymethyl)benzonitrile Hemi-(+)-di-p-toluoyltartrate |
| CAS | 128173-53-5 |
| Analyt- eller komponentnavne | 4-((1S)-4-(Dimethylamino)-1-(4-fluorophenyl)-1-hydroxybutyl)-3-(hydroxymethyl)benzonitrile Hemi-(+)-di-p-toluoyltartrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C60H64F2N4O12 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Butanedioic acid, 2,3-bis[(4-methylbenzoyl)oxy]-, (2S,3S)-, compd. with 4-[(1S)-4-(dimethylamino)-1-(4-fluorophenyl)-1-hydroxybutyl]-3-(hydroxymethyl)benzonitrile (1:2), Benzonitrile, 4-[4-(dimethylamino)-1-(4-fluorophenyl)-1-hydroxybutyl]-3-(hydroxymethyl)-, (S)-, (2S,3S)-2,3-bis[(4-methylbenzoyl)oxy]butanedioate (2:1) (salt) (9CI), Escitalopram Related Compound A as Hemi-di-toluoyltartrate, Escitalopram Oxalate Impurity E as (S)-Enantiomer Ditoluoyltartrate, Escitalopram Impurity E as (S)-Enantiomer Ditoluoyltartrate |
| InChI formel | InChI=1S/2C20H23FN2O2.C20H18O8/c2*1-23(2)11-3-10-20(25,17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24;1-11-3-7-13(8-4-11)19(25)27-15(17(21)22)16(18(23)24)28-20(26)14-9-5-12(2)6-10-14/h2*4-9,12,24-25H,3,10-11,14H2,1-2H3;3-10,15-16H,1-2H3,(H,21,22)(H,23,24)/t2*20-;15-,16-/m000/s1 |
| SMIL | CC1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=C(C=C2)C)C(=O)O.CN(C)CCCC(C1=CC=C(C=C1)F)(C2=C(C=C(C=C2)C#N)CO)O.CN(C)CCCC(C1=CC=C(C=C1)F)(C2=C(C=C(C=C2)C#N)CO)O |
| IUPAC navn | (2S,3S)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;4-[(1S)-4-(dimethylamino)-1-(4-fluorophenyl)-1-hydroxybutyl]-3-(hydroxymethyl)benzonitrile |
| Vis mere |