missing translation for 'onlineSavingsMsg'
Learn More

4-(1h-Imidazol-1-ylmethyl)benzaldehyde, 97%, Thermo Scientific™

Catalog Number p-4510658
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
102432-03-1
C11H10N2O
186.214
PYVCQHSLEIAQET-UHFFFAOYSA-N
11126983
4-(imidazol-1-ylmethyl)benzaldehyde
C1=CC(=CC=C1CN2C=CN=C2)C=O
This item is not returnable. View return policy
102432-03-1
C11H10N2O
186.214
PYVCQHSLEIAQET-UHFFFAOYSA-N
11126983
4-(imidazol-1-ylmethyl)benzaldehyde
C1=CC(=CC=C1CN2C=CN=C2)C=O
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-1h-imidazol-1-ylmethyl benzaldehyde, 4-imidazol-1-ylmethyl benzaldehyde, 4-1h-imidazol-1-yl methyl benzaldehyde, benzaldehyde,4-1h-imidazol-1-ylmethyl, acmc-20e4oe, 4-imidazolylmethyl benzaldehyde, benzaldehyde, 4-1h-imidazol-1-ylmethyl
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4-(1H-Imidazol-1-ylmethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'