missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
4-1BB Ligand/TNFSF9 Antibody (TKS-1), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81115UV
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
4-1BB Ligand/TNFSF9 Monoclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Flow Cytometry, ELISA.
Tekniske data
| 4-1BB Ligand/TNFSF9 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| TNFSF9 | |
| Murine B-cell lymphoma 2PK-3 cells injected into SD rats. | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins | |
| 8744 | |
| Mouse | |
| Purified |
| Flow Cytometry, ELISA | |
| TKS-1 | |
| Flow Cytometry, ELISA | |
| 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody binds to murine 4-1BB Ligand/TNFSF9 and competes with binding of 4-1BB. | |
| Store at 4°C in the dark. | |
| IgG κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion