missing translation for 'onlineSavingsMsg'
Få mere at vide

4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid (N2-Tritylolmesartan), Mikromol™

Artikelnummer. 30051349
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051349 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30051349 Leverandør Mikromol Leverandørnr. MM1317.310025

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid (N2-Tritylolmesartan)
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale N2-Tritylolmesartan
CAS 752179-89-8
Analyt- eller komponentnavne N2-Tritylolmesartan
Forsendelsestilstand Room Temperature
Molekylær formel C43 H40 N6 O3
Certificeringer/compliance ISO 9001
Synonym 1H-Imidazole-5-carboxylic acid, 4-(1-hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-, 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic acid
InChI formel InChI=1S/C43H40N6O3/c1-4-16-37-44-39(42(2,3)52)38(41(50)51)48(37)29-30-25-27-31(28-26-30)35-23-14-15-24-36(35)40-45-47-49(46-40)43(32-17-8-5-9-18-32,33-19-10-6-11-20-33)34-21-12-7-13-22-34/h5-15,17-28,52H,4,16,29H2,1-3H3,(H,50,51)
SMIL CCCc1nc(c(C(=O)O)n1Cc2ccc(cc2)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7)C(C)(C)O
IUPAC navn 5-(1-hydroxy-1-methyl-ethyl)-2-propyl-3-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylic acid
Molekylvægt (g/mol) 688.82
Mængde 25 mg
Formel vægt 688.3162 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.