Få mere at vide
4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazole-5-carboxylic Acid (Olmesartan), Mikromol™
Artikelnummer. 30462961
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462961 4 x 25 mg
30052501 25 mg
17919594 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30462961 Leverandør Mikromol Leverandørnr. MM1317.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazole-5-carboxylic Acid (Olmesartan), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Olmesartan Acid
CAS 144689-24-7
Analyt- eller komponentnavne Olmesartan Acid
Forsendelsestilstand Room Temperature
Molekylær formel C24 H26 N6 O3
Certificeringer/compliance ISO 9001
Synonym Olmesartan Medoxomil Imp. A (EP), 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazole-5-carboxylic Acid, Olmesartan, Olmesartan Acid, Olmesartan Medoxomil Impurity A
InChI formel InChI=1S/C24H26N6O3/c1-4-7-19-25-21(24(2,3)33)20(23(31)32)30(19)14-15-10-12-16(13-11-15)17-8-5-6-9-18(17)22-26-28-29-27-22/h5-6,8-13,33H,4,7,14H2,1-3H3,(H,31,32)(H,26,27,28,29)
Løsningstype Neat
SMIL CCCc1nc(c(C(=O)O)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4)C(C)(C)O
IUPAC navn 5-(2-hydroxypropan-2-yl)-2-propyl-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylic acid
Molekylvægt (g/mol) 446.5
Mængde 4 x 25 mg
Formel vægt 446.2066 g/mol