Få mere at vide
4-(1-Cyanoethyl)-1,2,3,4-tetrahydronaphthalene-1-carbonitrile, TRC
Artikelnummer. 17951415
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17951415 1 mg
17964125 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17951415 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9795451MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57964-39-3
Molekylær formel C14 H14 N2
Molekylvægt (g/mol) 210.27
Synonym 4-(1-Cyanoethyl)-1,2,3,4-tetrahydronaphthalene-1-carbonitrile, 4-Cyano-1,2,3,4-tetrahydro-α-methyl-1-naphthaleneacetonitrile, 1-Cyano-4-(1-cyanoethyl)-1,2,3,4-tetrahydronaphthalene
IUPAC navn 4-(1-cyanoethyl)tetralin-1-carbonitrile
SMIL CC(C#N)C1CCC(C#N)c2ccccc12
Tekniske data
Kemisk navn eller materiale 4-(1-Cyanoethyl)tetralin-1-carbonitrile
Mængde 1 mg
InChI formel InChI=1S/C14H14N2/c1-10(8-15)12-7-6-11(9-16)13-4-2-3-5-14(12)13/h2-5,10-12H,6-7H2,1H3
Formel vægt 210.1157