missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1,3-Thiazol-2-yl)benzaldehyde, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 95% |
Quantity | 250 mg |
Synonym | 4-thiazol-2-yl-benzaldehyde, 4-1,3-thiazol-2-yl benzaldehyde, 4-thiazol-2-yl benzaldehyde, benzaldehyde,4-2-thiazolyl, benzaldehyde, 4-2-thiazolyl, 4-2-thiazolyl benzaldehyde, 4-thiazol-2-ylbenzaldehyde, 4-2-thiazolyl-benzaldehyde |
Formula Weight | 189.24 |
Percent Purity | 95% |
Chemical Name or Material | 4-(1, 3-Thiazol-2-yl)benzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'