missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Oxazol-5-yl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-2700396
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
337508-66-4
C9H6ClNO3S
243.66
MFCD02681962
QCGUTLAPXWBIMM-UHFFFAOYSA-N
4-1,3-oxazol-5-yl benzenesulfonyl chloride, 4-oxazol-5-yl benzene-1-sulfonyl chloride, 4-oxazol-5-yl-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-5-oxazolyl, 4-1,3-oxazol-5-yl benzenesulfonylchloride, 4-1,3-oxazol-5-yl benzene-1-sulfonyl chloride, 4-1,3-oxazol-5-yl benzenesulphonyl chloride, 4-oxazol-5-ylbenzenesulfonyl-chloride, 4-oxazol-5-ylbenzene-sulfonyl-chloride, chloro 4-1,3-oxazol-5-yl phenyl sulfone
2776415
4-(1,3-oxazol-5-yl)benzenesulfonyl chloride
ClS(=O)(=O)C1=CC=C(C=C1)C1=CN=CO1
This item is not returnable. View return policy
337508-66-4
C9H6ClNO3S
243.66
MFCD02681962
QCGUTLAPXWBIMM-UHFFFAOYSA-N
4-1,3-oxazol-5-yl benzenesulfonyl chloride, 4-oxazol-5-yl benzene-1-sulfonyl chloride, 4-oxazol-5-yl-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-5-oxazolyl, 4-1,3-oxazol-5-yl benzenesulfonylchloride, 4-1,3-oxazol-5-yl benzene-1-sulfonyl chloride, 4-1,3-oxazol-5-yl benzenesulphonyl chloride, 4-oxazol-5-ylbenzenesulfonyl-chloride, 4-oxazol-5-ylbenzene-sulfonyl-chloride, chloro 4-1,3-oxazol-5-yl phenyl sulfone
2776415
4-(1,3-oxazol-5-yl)benzenesulfonyl chloride
ClS(=O)(=O)C1=CC=C(C=C1)C1=CN=CO1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 243.67
Percent Purity 97%
Chemical Name or Material 4-(1, 3-Oxazol-5-yl)benzenesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'