missing translation for 'onlineSavingsMsg'
Få mere at vide

[4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetic Acid, Mikromol™

Artikelnummer. 17903506
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17903506 25 mg
17902706 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17903506 Leverandør Mikromol Leverandørnr. MM0177.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetic Acid
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl)acetic Acid
CAS 55699-12-2
Analyt- eller komponentnavne (4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl)acetic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C14 H20 O3
Certificeringer/compliance ISO 9001
Synonym Oxymetazoline Hydrochloride Imp. D (EP), Oxymetazoline Imp. D (EP), [4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetic Acid, Oxymetazoline Hydrochloride Impurity D, Oxymetazoline Impurity D
InChI formel InChI=1S/C14H20O3/c1-8-6-11(14(3,4)5)13(17)9(2)10(8)7-12(15)16/h6,17H,7H2,1-5H3,(H,15,16)
SMIL Cc1cc(c(O)c(C)c1CC(=O)O)C(C)(C)C
IUPAC navn 2-(4-tert-butyl-3-hydroxy-2,6-dimethylphenyl)acetic acid
Molekylvægt (g/mol) 236.31
Mængde 25 mg
Formel vægt 236.1412 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.