Få mere at vide
[4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetic Acid, Mikromol™
Artikelnummer. 17902706
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17902706 100 mg
17903506 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17902706 Leverandør Mikromol Leverandørnr. MM0177.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
[4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl)acetic Acid
CAS 55699-12-2
Analyt- eller komponentnavne (4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl)acetic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C14 H20 O3
Certificeringer/compliance ISO 9001
Synonym Oxymetazoline Hydrochloride Imp. D (EP), Oxymetazoline Imp. D (EP), [4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetic Acid, Oxymetazoline Hydrochloride Impurity D, Oxymetazoline Impurity D
InChI formel InChI=1S/C14H20O3/c1-8-6-11(14(3,4)5)13(17)9(2)10(8)7-12(15)16/h6,17H,7H2,1-5H3,(H,15,16)
SMIL Cc1cc(c(O)c(C)c1CC(=O)O)C(C)(C)C
IUPAC navn 2-(4-tert-butyl-3-hydroxy-2,6-dimethylphenyl)acetic acid
Molekylvægt (g/mol) 236.31
Mængde 100 mg
Formel vægt 236.1412 g/mol