Få mere at vide
[4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]acetonitrile, Mikromol™
Artikelnummer. 17947714
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17947714 100 mg
17979544 25 mg
30461553 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17947714 Leverandør Mikromol Leverandørnr. MM0089.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
[4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]acetonitrile
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4-(1,1-Dimethylethyl)-2,6-dimethylphenyl)acetonitrile
CAS 84803-57-6
Analyt- eller komponentnavne (4-(1,1-Dimethylethyl)-2,6-dimethylphenyl)acetonitrile
Forsendelsestilstand Room Temperature
Molekylær formel C14 H19 N
Certificeringer/compliance ISO 17025
Synonym Xylometazoline Hydrochloride Imp. C (EP), Xylometazoline Imp. C (EP), [4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]acetonitrile, Xylometazoline Hydrochloride Impurity C, Xylometazoline Impurity C
InChI formel InChI=1S/C14H19N/c1-10-8-12(14(3,4)5)9-11(2)13(10)6-7-15/h8-9H,6H2,1-5H3
SMIL Cc1cc(cc(C)c1CC#N)C(C)(C)C
IUPAC navn 2-(4-tert-butyl-2,6-dimethylphenyl)acetonitrile
Molekylvægt (g/mol) 201.31
Mængde 100 mg
Formel vægt 201.1517 g/mol