Få mere at vide
(3S)-6,7-Dimethoxy-3-[(1R)-1,2,3,4-tetrahydro-6,7-dihydroxy-8-methoxy-2-methyl-1-isoquinolinyl]-1(3H)-isobenzofuranone, LoGiCal
Artikelnummer. 30461054
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30461054 10 mg
1 options
Artikelnummer. 30461054 Leverandør LoGiCal Leverandørnr. LGCFOR1232.02
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (3S)-6,7-Dimethoxy-3-[(1R)-1,2,3,4-tetrahydro-6,7-dihydroxy-8-methoxy-2-methyl-1-isoquinolinyl]-1(3H)-isobenzofuranone
CAS 78213-27-1
Analyt- eller komponentnavne (3S)-6,7-Dimethoxy-3-[(1R)-1,2,3,4-tetrahydro-6,7-dihydroxy-8-methoxy-2-methyl-1-isoquinolinyl]-1(3H)-isobenzofuranone
Forsendelsestilstand Room Temperature
Molekylær formel C21 H23 N O7
Certificeringer/compliance ISO 9001
Synonym (3S)-6,7-Dimethoxy-3-[(1R)-1,2,3,4-tetrahydro-6,7-dihydroxy-8-methoxy-2-methyl-1-isoquinolinyl]-1(3H)-Isobenzofuranone, 1(3H)-Isobenzofuranone, 6,7-dimethoxy-3-(1,2,3,4-tetrahydro-6,7-dihydroxy-8-methoxy-2-methyl-1-isoquinolinyl)-, [S-(R*,S*)]- (9CI)
InChI formel InChI=1S/C21H23NO7/c1-22-8-7-10-9-12(23)17(24)20(28-4)14(10)16(22)18-11-5-6-13(26-2)19(27-3)15(11)21(25)29-18/h5-6,9,16,18,23-24H,7-8H2,1-4H3/t16-,18+/m1/s1
Løsningstype Neat
SMIL COc1ccc2[C@H](OC(=O)c2c1OC)[C@@H]3N(C)CCc4cc(O)c(O)c(OC)c34
IUPAC navn (3S)-3-[(1R)-6,7-dihydroxy-8-methoxy-2-methyl-3,4-dihydro-1H-isoquinolin-1-yl]-6,7-dimethoxy-3H-2-benzofuran-1-one
Industry Type Clinical Testing
Molekylvægt (g/mol) 401.41
Mængde 10 mg
Formel vægt 401.1475 g/mol