Få mere at vide
(3S,5S)-3,5-Bis(tert-butyldimethylsilyloxy)-2-methyl-1-cyclohexen-1-ol 1-Trifluoromethanesulfonate, TRC
Artikelnummer. 30337082
Skift visning
Mængde:
2 mg
2.5 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30337082 2.5 mg 2.5mg
30341321 2 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30337082 Leverandør Toronto Research Chemicals Leverandørnr. TRCB4157952.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (3S,5S)-3,5-Bis(tert-butyldimethylsilyloxy)-2-methyl-1-cyclohexen-1-ol 1-Trifluoromethanesulfonate
CAS 189894-13-1
Molekylær formel C20H39F3O5SSi2
Mængde 2.5 mg
Synonym 1,1,1-Trifluoro-methanesulfonic Acid (3S,5S)-3,5-Bis[[(1,1-dimethylethyl)dimethylsilyl]oxy]-2-methyl-1-cyclohexen-1-yl Ester
InChI formel InChI=1S/C20H39F3O5SSi2/c1-14-16(26-29(24,25)20(21,22)23)12-15(27-30(8,9)18(2,3)4)13-17(14)28-31(10,11)19(5,6)7/h15,17H,12-13H2,1-11H3/t15-,17+/m1/s1
SMIL CC1=C(C[C@H](C[C@@H]1O[Si](C)(C)C(C)(C)C)O[Si](C)(C)C(C)(C)C)OS(=O)(=O)C(F)(F)F
IUPAC navn [(3S,5S)-3,5-bis[[tert-butyl(dimethyl)silyl]oxy]-2-methylcyclohexen-1-yl] trifluoromethanesulfonate
Molekylvægt (g/mol) 504.75
Formel vægt 504.2009