Få mere at vide
(3S,4R)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-phenylpiperidine Hydrochloride (Desfluoroparoxetine Hydrochloride), Mikromol™
Beskrivelse
(3S,4R)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-phenylpiperidine Hydrochloride (Desfluoroparoxetine Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (3S,4R)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-phenylpiperidine Hydrochloride (Desfluoroparoxetine Hydrochloride) |
| CAS | 1394842-91-1 |
| Analyt- eller komponentnavne | (3S,4R)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-phenylpiperidine Hydrochloride (Desfluoroparoxetine Hydrochloride) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H21 N O3 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Paroxetine Hydrochloride Anhydrous Imp. A (EP), Paroxetine Imp. A (EP), (3S,4R)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-phenylpiperidine Hydrochloride, Paroxetine Hydrochloride Anhydrous Imp. A (EP) as Hydrochloride, Desfluoroparoxetine Hydrochloride, Paroxetine Related Compound B as Hydrochloride, Paroxetine Hydrochloride Hemihydrate Impurity A as Hydrochloride, Paroxetine Hydrochloride Impurity A as Hydrochloride, Paroxetine Impurity A as Hydrochloride, Paroxetine Impurity A as Hydrochloride |
| InChI formel | InChI=1S/C19H21NO3.ClH/c1-2-4-14(5-3-1)17-8-9-20-11-15(17)12-21-16-6-7-18-19(10-16)23-13-22-18;/h1-7,10,15,17,20H,8-9,11-13H2;1H/t15-,17-;/m0./s1 |
| SMIL | Cl.C(Oc1ccc2OCOc2c1)[C@@H]3CNCC[C@H]3c4ccccc4 |
| IUPAC navn | (3S,4R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-phenylpiperidine;hydrochloride |
| Vis mere |