Få mere at vide
(3S)-1,3-Bis-O-tert-Butyldimethylsilyl 3-Hydroxy Vitamin D2 SO2 Adduct (Mixture of Diastereomers), TRC
Artikelnummer. 30331654
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30331654 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331654 Leverandør Toronto Research Chemicals Leverandørnr. TRCB4158405MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (3S)-1,3-Bis-O-tert-Butyldimethylsilyl 3-Hydroxy Vitamin D2 SO2 Adduct (Mixture of Diastereomers)
CAS 170081-46-6
Molekylær formel C40H72O4SSi2
Mængde 5 mg
Synonym [[(1alpha,3beta,7E,22E)-6,19-Sulfonyl-9,10-secoergosta-5(10),7,22-triene-1,3-diyl]bis(oxy)]bis[(1,1-dimethylethyl)dimethyl-silane
InChI formel InChI=1S/C40H72O4SSi2/c1-27(2)28(3)18-19-29(4)34-20-21-35-30(17-16-22-40(34,35)11)23-37-32-24-31(43-46(12,13)38(5,6)7)25-36(33(32)26-45(37,41)42)44-47(14,15)39(8,9)10/h18-19,23,27-29,31,34-37H,16-17,20-22,24-26H2,1-15H3/b19-18+,30-23+/t28-,29?,31+,34+,35-,36-,37?,40+/m0/s1
SMIL CC(C)[C@@H](C)\C=C\C(C)[C@H]1CC[C@H]2\C(=C\C3C4=C(CS3(=O)=O)[C@H](C[C@@H](C4)O[Si](C)(C)C(C)(C)C)O[Si](C)(C)C(C)(C)C)\CCC[C@]12C
IUPAC navn [(4S,6R)-1-[(E)-[(1R,3aS,7aR)-1-[(E,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]methyl]-6-[tert-butyl(dimethyl)silyl]oxy-2,2-dioxo-1,3,4,5,6,7-hexahydro-2-benzothiophen-4-yl]oxy-tert-butyl-dimethylsilane
Molekylvægt (g/mol) 705.23
Formel vægt 704.469