Få mere at vide
(3S)-1,3-Bis-O-(tert-Butyldimethylsilyl)-3-hydroxy-5,6-trans-vitamin D2, TRC
Artikelnummer. 30337046
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30337046 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30337046 Leverandør Toronto Research Chemicals Leverandørnr. TRCB691910100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (3S)-1,3-Bis-O-(tert-Butyldimethylsilyl)-3-hydroxy-5,6-trans-vitamin D2
CAS 111594-58-2
Molekylær formel C40H72O2Si2
Mængde 100 mg
Synonym [[(1alpha,3beta,5E,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraene-1,3-diyl]bis(oxy)]bis[(1,1-dimethylethyl)dimethyl-silane
InChI formel InChI=1S/C40H72O2Si2/c1-28(2)29(3)19-20-30(4)35-23-24-36-32(18-17-25-40(35,36)12)21-22-33-26-34(41-43(13,14)38(6,7)8)27-37(31(33)5)42-44(15,16)39(9,10)11/h19-22,28-30,34-37H,5,17-18,23-27H2,1-4,6-16H3/b20-19+,32-21+,33-22+/t29-,30+,34+,35+,36-,37-,40+/m0/s1
SMIL CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C\3/C[C@H](C[C@H](O[Si](C)(C)C(C)(C)C)C3=C)O[Si](C)(C)C(C)(C)C)\CCC[C@]12C
IUPAC navn [(1S,3E,5R)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-5-[tert-butyl(dimethyl)silyl]oxy-2-methylidenecyclohexyl]oxy-tert-butyl-dimethylsilane
Molekylvægt (g/mol) 641.17
Formel vægt 640.5071