Learn More
(3S)-1,2,3,4-Tetrahydro-6,7-dimethoxy-3-isoquinolinecarboxylic Acid Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (3S)-1,2,3,4-Tetrahydro-6,7-dimethoxy-3-isoquinolinecarboxylic acid hydrochloride |
| CAS | 82586-62-7 |
| Molecular Formula | C12 H15 N O4 . Cl H |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C12H15NO4.ClH/c1-16-10-4-7-3-9(12(14)15)13-6-8(7)5-11(10)17-2;/h4-5,9,13H,3,6H2,1-2H3,(H,14,15);1H/t9-;/m0./s1 |
| SMILES | Cl.COc1cc2CN[C@@H](Cc2cc1OC)C(=O)O |
| IUPAC Name | (3S)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid;hydrochloride |
| Molecular Weight (g/mol) | 273.713 |
| Formula Weight | 273.077 |