Få mere at vide
(3RS)-N-Methyl-3-phenyl-3-[2-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride (2-Trifluoromethylisomer of Fluoxetine Hydrochloride), Mikromol™
Artikelnummer. 30462565
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462565 4 x 25 mg
17942196 100 mg
17951446 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30462565 Leverandør Mikromol Leverandørnr. MM0256.1700254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(3RS)-N-Methyl-3-phenyl-3-[2-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride (2-Trifluoromethylisomer of Fluoxetine Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (3RS)-N-Methyl-3-phenyl-3-[2-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride
CAS 2243504-22-3
Analyt- eller komponentnavne (3RS)-N-Methyl-3-phenyl-3-[2-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C17 H18 F3 N O . Cl H
Certificeringer/compliance ISO 17025
Synonym (3RS)-N-Methyl-3-phenyl-3-[2-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride, 2-Trifluoromethylisomer of Fluoxetine Hydrochloride
InChI formel InChI=1S/C17H18F3NO.ClH/c1-21-12-11-15(13-7-3-2-4-8-13)22-16-10-6-5-9-14(16)17(18,19)20;/h2-10,15,21H,11-12H2,1H3;1H
Løsningstype Neat
SMIL Cl.CNCCC(Oc1ccccc1C(F)(F)F)c2ccccc2
IUPAC navn N-methyl-3-phenyl-3-[2-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
Molekylvægt (g/mol) 345.79
Mængde 4 x 25 mg
Formel vægt 345.1107 g/mol