Få mere at vide
(3RS)-3-[(2RS)-(2-Cyclopentyl-2-hydroxy-2-phenylacetyl)oxy]-1,1-dimethylpyrrolidinium Bromide, Mikromol™
Beskrivelse
(3RS)-3-[(2RS)-(2-Cyclopentyl-2-hydroxy-2-phenylacetyl)oxy]-1,1-dimethylpyrrolidinium Bromide
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (3RS)-3-[(2RS)-(2-Cyclopentyl-2-hydroxy-2-phenylacetyl)oxy]-1,1-dimethylpyrrolidinium Bromide |
| CAS | 58493-54-2 |
| Analyt- eller komponentnavne | (3RS)-3-[(2RS)-(2-Cyclopentyl-2-hydroxy-2-phenylacetyl)oxy]-1,1-dimethylpyrrolidinium Bromide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H28 N O3 . Br |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Pyrrolidinium, 3-[[(2R)-2-cyclopentyl-2-hydroxy-2-phenylacetyl]oxy]-1,1-dimethyl-, bromide (1:1), (3R)-rel-, Pyrrolidinium, 3-[(cyclopentylhydroxyphenylacetyl)oxy]-1,1-dimethyl-, bromide, (R*,R*)- (9CI), Ritropirronium bromide, threo-Glycopyrronium bromide, Glycopyrrolate Erythro Isomer ((RS)-3-[(RS)-2-cyclopentyl-2-hydroxy-2-phenylacetoxy]-1,1-dimethylpyrrolidinium bromide), Glycopyrrolate Erythro Isomer (USP), Glycopyrronium Bromide Imp. N (EP), Pyrrolidinium, 3-[(cyclopentylhydroxyphenylacetyl)oxy]-1,1-dimethyl-, bromide, (R*,R*)-(+/-)-, Glycopyrronium Bromide Impurity N as Bromide, Glycopyrronium Impurity N as Bromide |
| InChI formel | InChI=1S/C19H28NO3.BrH/c1-20(2)13-12-17(14-20)23-18(21)19(22,16-10-6-7-11-16)15-8-4-3-5-9-15;/h3-5,8-9,16-17,22H,6-7,10-14H2,1-2H3;1H/q+1;/p-1/t17-,19+;/m1./s1 |
| SMIL | [Br-].C[N+]1(C)CC[C@H](C1)OC(=O)[C@@](O)(C2CCCC2)c3ccccc3 |
| IUPAC navn | [(3R)-1,1-dimethylpyrrolidin-1-ium-3-yl] (2R)-2-cyclopentyl-2-hydroxy-2-phenylacetate;bromide |
| Vis mere |