Få mere at vide
(3R-trans)-[3,5-Bis[[(1,1-dimethylethyl)dimethylsilyl]oxy]cyclohexylidene]-acetic Acid Ethyl Ester, TRC
Artikelnummer. 30346488
Skift visning
Mængde:
2 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30346488 5 mg
30332106 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30346488 Leverandør Toronto Research Chemicals Leverandørnr. TRCB4298055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (3R-trans)-(3,5-Bis(((1,1-dimethylethyl)dimethylsilyl)oxy)cyclohexylidene)-acetic Acid Ethyl Ester
CAS 139356-36-8
Molekylær formel C22H44O4Si2
Mængde 5 mg
Synonym 2-[(3R,5R)-3,5-Bis[[(1,1-dimethylethyl)dimethylsilyl]oxy]cyclohexylidene]-acetic Acid Ethyl Ester, [(3R,5R)-3,5-bis[[(1,1-dimethylethyl)dimethylsilyl]oxy]cyclohexylidene]-acetic Acid Ethyl Ester
InChI formel InChI=1S/C22H44O4Si2/c1-12-24-20(23)15-17-13-18(25-27(8,9)21(2,3)4)16-19(14-17)26-28(10,11)22(5,6)7/h15,18-19H,12-14,16H2,1-11H3/t18-,19-/m1/s1
SMIL CCOC(=O)C=C1C[C@H](C[C@@H](C1)O[Si](C)(C)C(C)(C)C)O[Si](C)(C)C(C)(C)C
IUPAC navn ethyl 2-[(3R,5R)-3,5-bis[[tert-butyl(dimethyl)silyl]oxy]cyclohexylidene]acetate
Molekylvægt (g/mol) 428.75
Formel vægt 428.2778