Få mere at vide
(3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethylbutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Ammonium Salt (Tenivastatin Ammonium Salt), Mikromol™
Beskrivelse
(3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethylbutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Ammonium Salt (Tenivastatin Ammonium Salt)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Simvastatin Hydroxy Acid, Ammonium Salt |
| CAS | 139893-43-9 |
| Analyt- eller komponentnavne | Simvastatin Hydroxy Acid, Ammonium Salt |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C25 H39 O6 . H4 N |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Simvastatin Imp. A (EP), Tenivastatin Calcium, 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-beta,delta-dihydroxy-2,6-dimethyl-, ammonium salt (1:1), (betaR,deltaR,1S,2S,6R,8S,8aR)-, 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-beta,delta-dihydroxy-2,6-dimethyl-, monoammonium salt, (betaR,deltaR,1S,2S,6R,8S,8aR)- (9CI), 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-beta,delta-dihydroxy-2,6-dimethyl-, monoammonium salt, [1S-[1alpha(betaS*,deltaS*),2alpha,6beta,8beta,8aalpha]]-, Ammonium simvastatin, Simvastatin acid ammonium salt, Simvastatin ammonium salt, Simvastatin carboxylic acid ammonium salt, Simvastatin Impurity A as Ammonium Salt |
| InChI formel | InChI=1S/C25H40O6.H3N/c1-6-25(4,5)24(30)31-21-12-15(2)11-17-8-7-16(3)20(23(17)21)10-9-18(26)13-19(27)14-22(28)29;/h7-8,11,15-16,18-21,23,26-27H,6,9-10,12-14H2,1-5H3,(H,28,29);1H3/t15-,16-,18+,19+,20-,21-,23-;/m0./s1 |
| SMIL | [NH4+].CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)[O-])[C@@H]12 |
| IUPAC navn | azanium;(3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-(2,2-dimethylbutanoyloxy)-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate |
| Vis mere |