Få mere at vide
(3R,4R)-3-Ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]dihydrofuran-2(3H)-one Hydrochloride (Isopilocarpine Hydrochloride), Mikromol™
Artikelnummer. 18081210
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18081210 25 mg
18031000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 18081210 Leverandør Mikromol Leverandørnr. MM0338.010025
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(3R,4R)-3-Ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]dihydrofuran-2(3H)-one Hydrochloride (Isopilocarpine Hydrochloride)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (3R,4R)-3-Ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]dihydrofuran-2(3H)-one Hydrochloride (Isopilocarpine Hydrochloride)
CAS 28958-85-2
Analyt- eller komponentnavne (3R,4R)-3-Ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]dihydrofuran-2(3H)-one Hydrochloride (Isopilocarpine Hydrochloride)
Forsendelsestilstand Room Temperature
Molekylær formel C11 H16 N2 O2 . Cl H
Certificeringer/compliance ISO 17025
Synonym Pilocarpine Hydrochloride Imp. A (EP), Pilocarpine Imp. A (EP), (3R,4R)-3-Ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]dihydrofuran-2(3H)-one Hydrochloride, Pilocarpine Hydrochloride Imp. A (EP) as Hydrochloride, Isopilocarpine Hydrochloride, Pilocarpine Hydrochloride Impurity A as Hydrochloride, Pilocarpine Nitrate Impurity A as Hydrochloride, Pilocarpine Impurity A as Hydrochloride, Pilocarpine Impurity A as Hydrochloride
InChI formel InChI=1S/C11H16N2O2.ClH/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;/h5,7-8,10H,3-4,6H2,1-2H3;1H/t8-,10+;/m0./s1
SMIL Cl.CC[C@@H]1[C@@H](Cc2cncn2C)COC1=O
IUPAC navn (3R,4R)-3-ethyl-4-[(3-methylimidazol-4-yl)methyl]oxolan-2-one;hydrochloride
Molekylvægt (g/mol) 244.72
Mængde 25 mg
Formel vægt 244.0979 g/mol