Få mere at vide
(3R,4R)-3-[(6-Amino-4-pyrimidinyl)methylamino]-4-methyl-Beta-oxo-1-piperidinepropanenitrile, TRC
Artikelnummer. 30137676
Skift visning
Mængde:
10 mg
25 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137676 25 mg
30134005 500 mg
30008117 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137676 Leverandør Toronto Research Chemicals Leverandørnr. TRCA59395025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1640971-60-3
Molekylær formel C14H20N6O
Molekylvægt (g/mol) 288.35
Synonym (3R,4R)-3-[(6-Amino-4-Pyrimidinyl)Methylamino]-4-Methyl-B-Oxo-1-Piperidinepropanenitrile
IUPAC navn 3-[(3R,4R)-3-[(6-aminopyrimidin-4-yl)-methylamino]-4-methylpiperidin-1-yl]-3-oxopropanenitrile
SMIL C[C@@H]1CCN(C[C@@H]1N(C)c2cc(N)ncn2)C(=O)CC#N
Tekniske data
Kemisk navn eller materiale (3R,4R)-3-[(6-Amino-4-pyrimidinyl)methylamino]-4-methyl-beta-oxo-1-piperidinepropanenitrile
Analyt- eller komponentnavne (3R,4R)-3-[(6-Amino-4-pyrimidinyl)methylamino]-4-methyl-beta-oxo-1-piperidinepropanenitrile
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C14H20N6O/c1-10-4-6-20(14(21)3-5-15)8-11(10)19(2)13-7-12(16)17-9-18-13/h7,9-11H,3-4,6,8H2,1-2H3,(H2,16,17,18)/t10-,11+/m1/s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 25 mg
Formel vægt 288.1699