Få mere at vide
[(3R)-3-[[(1R)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic Acid Hydrochloride (ent-Benazepril Hydrochloride), Mikromol
Beskrivelse
[(3R)-3-[[(1R)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic Acid Hydrochloride (ent-Benazepril Hydrochloride), Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (3R)-3-[[(1R)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid, monohydrochloride |
| CAS | 215447-89-5 |
| Analyt- eller komponentnavne | (3R)-3-[[(1R)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid, monohydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C24 H28 N2 O5 . H Cl |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1H-1-Benzazepine-1-acetic acid, 3-[[(1R)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-, hydrochloride (1:1), (3R)-, 1H-1-Benzazepine-1-acetic acid, 3-[[(1R)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-, monohydrochloride, (3R)- (9CI), (3R)-3-[[(1R)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid hydrochloride (1:1), Benazepril Hydrochloride Imp. A (EP) as Hydrochloride |
| InChI formel | InChI=1S/C24H28N2O5.ClH/c1-2-31-24(30)20(14-12-17-8-4-3-5-9-17)25-19-15-13-18-10-6-7-11-21(18)26(23(19)29)16-22(27)28;/h3-11,19-20,25H,2,12-16H2,1H3,(H,27,28);1H/t19-,20-;/m1./s1 |
| Løsningstype | Neat |
| SMIL | Cl.CCOC(=O)[C@@H](CCc1ccccc1)N[C@@H]2CCc3ccccc3N(CC(=O)O)C2=O |
| Vis mere |