Få mere at vide
3E,8Z-Tetradecadienyl Acetate (>80%), TRC
Artikelnummer. 30009735
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009735 1 g
30007412 500 mg
30003641 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009735 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2921101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 163041-87-0
Molekylær formel C16H28O2
Molekylvægt (g/mol) 252.39
Synonym (E,Z)-3,8-Tetradecadienyl Acetate, trans-3,cis-8-Tetradecadienyl Acetate, (E,Z)-3,8-Tetradecadien-1-ol Acetate, (3E,8Z)-3,8-Tetradecadien-1-ol Acetate, (3E,8Z)-3,8-Tetradecadien-1-ol 1-Acetate;
IUPAC navn [(3E,8Z)-tetradeca-3,8-dienyl] acetate
SMIL CCCCC\C=C/CCC\C=C\CCOC(=O)C
Tekniske data
Kemisk navn eller materiale 3E,8Z-Tetradecadienyl Acetate
Mængde 1 g
InChI formel InChI=1S/C16H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-18-16(2)17/h7-8,12-13H,3-6,9-11,14-15H2,1-2H3/b8-7-,13-12+
Formel vægt 252.2089