Få mere at vide
3Beta-tert-Butyldimethylsilyloxy-17-methyl-5Alpha-androstane-17Beta-ol-d5, TRC
Artikelnummer. 30335381
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30335381 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30335381 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6912125MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3beta-tert-Butyldimethylsilyloxy-17-methyl-5alpha-androstane-17beta-ol-d5
CAS 853904-64-0
Molekylær formel C26H43D5O2Si
Mængde 5 mg
Synonym (3beta,5alpha,17beta)-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-17-(methyl-d3)-androstan-16,16-d2-17-ol
InChI formel InChI=1S/C26H48O2Si/c1-23(2,3)29(7,8)28-19-11-14-24(4)18(17-19)9-10-20-21(24)12-15-25(5)22(20)13-16-26(25,6)27/h18-22,27H,9-17H2,1-8H3/t18-,19-,20+,21-,22-,24-,25-,26-/m0/s1/i6D3,16D2
SMIL [2H]C([2H])([2H])[C@]1(O)C([2H])([2H])C[C@H]2[C@@H]3CC[C@H]4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)O[Si](C)(C)C(C)(C)C
IUPAC navn (3S,5S,8R,9S,10S,13S,14S,17S)-3-[tert-butyl(dimethyl)silyl]oxy-16,16-dideuterio-10,13-dimethyl-17-(trideuteriomethyl)-2,3,4,5,6,7,8,9,11,12,14,15-dodecahydro-1H-cyclopenta[a]phenanthren-17-ol
Molekylvægt (g/mol) 425.77
Formel vægt 425.3737