Få mere at vide
3beta,5alpha-Tetrahydrodeoxycorticosterone, TRC
Artikelnummer. 17990675
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17990675 25 mg
17930695 5 mg
17901715 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17990675 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29329025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 567-01-1
Molekylær formel C21 H34 O3
Molekylvægt (g/mol) 334.49
IUPAC navn 2-hydroxy-1-[(3S,5S,8R,9S,10S,13S,14S,17S)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
SMIL C[C@]12CC[C@H](O)C[C@@H]1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@@]4(C)CC[C@H]23
Tekniske data
Kemisk navn eller materiale 3beta,5alpha-Tetrahydrodeoxycorticosterone
Mængde 25 mg
Synonym Pregnan-20-one, 3,21-dihydroxy-, (3β,5α)-, 5α-Pregnan-20-one, 3β,21-dihydroxy- (8CI), (3β,5α)-3,21-Dihydroxypregnan-20-one, 3β,21-Dihydroxy-5α-pregnan-20-one, 3β,5α-Tetrahydrodeoxycorticosterone, (3β,5α)- 3,21-Dihydroxypregnan-20-one, Epiallopregnanolone
InChI formel InChI=1S/C21H34O3/c1-20-9-7-14(23)11-13(20)3-4-15-16-5-6-18(19(24)12-22)21(16,2)10-8-17(15)20/h13-18,22-23H,3-12H2,1-2H3/t13-,14-,15-,16-,17-,18+,20-,21-/m0/s1
Formel vægt 334.2508