Få mere at vide
(3Beta,5Alpha)-Cholesta-8,14-dien-3-ol, TRC
Artikelnummer. 30362181
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362181 50 mg
30360799 500 mg
30353648 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30362181 Leverandør Toronto Research Chemicals Leverandørnr. TRCC43152550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19431-20-0
Molekylær formel C27H44O
Molekylvægt (g/mol) 384.64
IUPAC navn (3S,5S,10S,13R,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,11,12,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-ol
SMIL CC(C)CCCC(C)[C@H]1CC=C2C3=C(CC[C@]12C)[C@@]4(C)CC[C@H](O)C[C@H]4CC3
Tekniske data
Kemisk navn eller materiale (3beta,5alpha)-Cholesta-8,14-dien-3-ol
Mængde 50 mg
InChI formel InChI=1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h12,18-21,23,28H,6-11,13-17H2,1-5H3/t19-,20+,21+,23-,26+,27-/m1/s1
Formel vægt 384.3392